Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf20l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081409.1 | NP_001074878.1 | 15 - 16 | 2216 | 64 | |
GenBank mRNA | AK040888.1 | - | 6 - 7 | 947 | 64 | |
AK041825.1 | - | 4 - 5 | 2485 | 64 | ||
AK053606.1 | - | 5 - 6 | 575 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |