Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scai | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178778.3 | NP_848893.2 | 9 - 10 | 950 | 74 | |
GenBank mRNA | AK044772.1 | - | 9 - 10 | 946 | 74 | |
AK082408.1 | - | 10 - 11 | 1052 | 74 | ||
AK083280.1 | - | 9 - 10 | 952 | 74 | ||
AK163712.1 | - | 9 - 10 | 954 | 74 | ||
BC132485.1 | - | 9 - 10 | 925 | 40130762 | 74 | |
BC144986.1 | - | 10 - 11 | 1026 | 74 | ||
BC144987.1 | - | 9 - 10 | 1001 | 74 | ||
BC144989.1 | - | 10 - 11 | 1026 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |