Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178884.5 | NP_849215.3 | 1 - 2 | 1241 | 87 | |
GenBank mRNA | AK029863.1 | - | 1 - 2 | 1031 | 87 | |
AK031074.1 | - | 1 - 2 | 1242 | 87 | ||
AK034852.1 | - | 1 - 2 | 1152 | 87 | ||
AK048144.1 | - | 1 - 2 | 432 | 87 | ||
AK162473.1 | - | 1 - 2 | 1151 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus 1700 Probe spans exons |