Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100007736.mm.1 |
NM_181405.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007736.mm.1 |
NM_181405.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007736.mm.1 |
TC0100000921.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100007730.mm.1 |
NM_181405.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007730.mm.1 |
NM_181405.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007730.mm.1 |
TC0100000921.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100007731.mm.1 |
NM_181405.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007731.mm.1 |
NM_181405.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007731.mm.1 |
TC0100000921.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100007729.mm.1 |
NM_181405.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007729.mm.1 |
NM_181405.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007729.mm.1 |
TC0100000921.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100007734.mm.1 |
NM_181405.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007734.mm.1 |
NM_181405.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007734.mm.1 |
TC0100000921.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100007732.mm.1 |
NM_181405.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007732.mm.1 |
NM_181405.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007732.mm.1 |
TC0100000921.mm.1 |
MTA TC |