Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173778.3 | NP_776139.1 | 1 - 2 | 150 | 70 | |
GenBank mRNA | AK030178.1 | - | 1 - 2 | 151 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |