Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000012528.mm.1 |
NM_175155.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012528.mm.1 |
NM_175155.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012528.mm.1 |
TC1000001716.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012535.mm.1 |
NM_175155.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012535.mm.1 |
NM_175155.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012535.mm.1 |
TC1000001716.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012524.mm.1 |
NM_175155.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012524.mm.1 |
NM_175155.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012524.mm.1 |
TC1000001716.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012526.mm.1 |
NM_175155.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012526.mm.1 |
NM_175155.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012526.mm.1 |
TC1000001716.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012534.mm.1 |
NM_175155.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012534.mm.1 |
NM_175155.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012534.mm.1 |
TC1000001716.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012531.mm.1 |
NM_175155.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012531.mm.1 |
NM_175155.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012531.mm.1 |
TC1000001716.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012525.mm.1 |
NM_175155.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012525.mm.1 |
NM_175155.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012525.mm.1 |
TC1000001716.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012527.mm.1 |
NM_175155.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012527.mm.1 |
NM_175155.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012527.mm.1 |
TC1000001716.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012529.mm.1 |
NM_175155.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012529.mm.1 |
NM_175155.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012529.mm.1 |
TC1000001716.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012532.mm.1 |
NM_175155.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012532.mm.1 |
NM_175155.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012532.mm.1 |
TC1000001716.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012533.mm.1 |
NM_175155.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012533.mm.1 |
NM_175155.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012533.mm.1 |
TC1000001716.mm.1 |
MTA TC |