Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtap7d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177293.3 | NP_796267.2 | 4 - 5 | 416 | 83 | |
GenBank mRNA | AK030211.1 | - | 3 - 4 | 333 | 83 | |
AK049164.1 | - | 4 - 5 | 406 | 83 | ||
BC096764.1 | - | 4 - 5 | 418 | 84 | ||
BC118963.2 | - | 3 - 4 | 333 | 40095274 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |