Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Umodl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177465.4 | NP_803416.2 | 17 - 18 | 3457 | 64 | |
GenBank mRNA | AK040446.1 | - | 9 - 10 | 1958 | 64 | |
AY771616.1 | - | 18 - 19 | 3374 | 64 | ||
AY771617.1 | - | 17 - 18 | 3461 | 64 | ||
AY771618.1 | - | 17 - 18 | 3116 | 64 | ||
BC051427.1 | - | 17 - 18 | 3112 | 64 | ||
BC132381.1 | - | 4 - 5 | 646 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |