Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177697.3 | NP_808365.2 | 18 - 19 | 2120 | 73 | |
GenBank mRNA | AK086942.1 | - | 17 - 18 | 1718 | 73 | |
AK132732.1 | - | 18 - 19 | 2121 | 73 | ||
AK136898.1 | - | 17 - 18 | 1719 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |