Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177765.3 | NP_808433.2 | 1 - 2 | 287 | 89 | |
GenBank mRNA | AK029565.1 | - | 1 - 2 | 288 | 89 | |
AK161519.1 | - | 1 - 2 | 109 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |