Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wee1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009516.3 | NP_033542.2 | 8 - 9 | 1759 | 104 | |
GenBank mRNA | AK011212.1 | - | 8 - 9 | 1535 | 104 | |
AK039572.1 | - | 8 - 9 | 1075 | 104 | ||
AK215663.1 | - | 3 - 4 | 238 | 104 | ||
BC006852.1 | - | 8 - 9 | 1778 | 3156953 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |