Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gosr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016810.3 | NP_058090.2 | 6 - 7 | 521 | 146 | |
GenBank mRNA | AF079901.1 | - | 6 - 7 | 544 | 146 | |
AK133836.1 | - | 6 - 7 | 531 | 146 | ||
AK134515.1 | - | 6 - 7 | 524 | 146 | ||
AK147799.1 | - | 6 - 7 | 545 | 146 | ||
AK154972.1 | - | 6 - 7 | 547 | 146 | ||
AK161386.1 | - | 6 - 7 | 535 | 146 | ||
BC008542.1 | - | 6 - 7 | 521 | 3495882 | 146 | |
BC051661.1 | - | 6 - 7 | 540 | 30007684 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |