Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK011899.1 | - | 4 - 5 | 489 | 148 | |
AK082940.1 | - | 4 - 5 | 509 | 148 | ||
AK167383.1 | - | 4 - 5 | 497 | 148 | ||
AK167848.1 | - | 4 - 5 | 493 | 148 | ||
AY082332.1 | - | 4 - 5 | 463 | 148 | ||
BC050206.1 | - | 4 - 5 | 471 | 30009463 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |