Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rprd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081293.1 | NP_001074762.1 | 8 - 9 | 1410 | 73 | |
GenBank mRNA | AK083674.1 | - | 8 - 9 | 1179 | 73 | |
AK087157.1 | - | 8 - 9 | 1182 | 73 | ||
AK129150.1 | - | 7 - 8 | 957 | 73 | ||
AK134893.1 | - | 1 - 2 | 267 | 73 | ||
AK154692.1 | - | 7 - 8 | 1084 | 73 | ||
BC067054.1 | - | 8 - 9 | 1215 | 30542928 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |