Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gas5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002840.2 | 2 - 3 | 2423 | 71 | ||
GenBank mRNA | AK083453.1 | - | 2 - 3 | 2424 | 71 | |
BC055883.1 | - | 6 - 7 | 934 | 71 | ||
BC080708.1 | - | 4 - 5 | 1480 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |