Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Echs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053119.2 | NP_444349.1 | 2 - 3 | 314 | 75 | |
GenBank mRNA | AK040391.1 | - | 2 - 3 | 315 | 75 | |
AK044954.1 | - | 2 - 3 | 309 | 75 | ||
AK086762.1 | - | 2 - 3 | 307 | 75 | ||
AK088018.1 | - | 2 - 3 | 300 | 75 | ||
AK167404.1 | - | 2 - 3 | 294 | 75 | ||
AK210204.1 | - | 1 - 2 | 126 | 75 | ||
BC002178.1 | - | 2 - 3 | 280 | 75 | ||
BC072658.1 | - | 2 - 3 | 317 | 30357710 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |