Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013256.2 | NP_001013274.2 | 8 - 9 | 1657 | 65 | |
NM_001143790.1 | NP_001137262.1 | 8 - 9 | 1657 | 65 | ||
NM_001281464.1 | NP_001268393.1 | 8 - 9 | 1528 | 65 | ||
NM_001281465.1 | NP_001268394.1 | 8 - 9 | 1641 | 65 | ||
GenBank mRNA | AK038651.1 | - | 8 - 9 | 1444 | 65 | |
AK044055.1 | - | 8 - 9 | 1436 | 65 | ||
AK145919.1 | - | 8 - 9 | 1472 | 65 | ||
AK165906.1 | - | 8 - 9 | 1457 | 65 | ||
AK216928.1 | - | 2 - 3 | 285 | 65 | ||
BC060629.1 | - | 4 - 5 | 707 | 6847619 | 65 | |
BC066078.1 | - | 8 - 9 | 1472 | 65 | ||
BC072625.1 | - | 8 - 9 | 1485 | 65 | ||
BC147495.1 | - | 8 - 9 | 1455 | 9056325 | 65 | |
BC147497.1 | - | 8 - 9 | 1455 | 9056354 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |