Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eci3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026947.4 | NP_081223.1 | 4 - 5 | 414 | 102 | |
GenBank mRNA | AK007578.1 | - | 4 - 5 | 415 | 102 | |
AK131590.1 | - | 4 - 5 | 415 | 102 | ||
BC014724.1 | - | 4 - 5 | 388 | 4219360 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |