Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200034625.mm.1 |
NM_001166597.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034625.mm.1 |
NM_001166597.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034625.mm.1 |
NM_138306.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034625.mm.1 |
NM_138306.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034625.mm.1 |
TC0200004130.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200034626.mm.1 |
NM_001166597.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034626.mm.1 |
NM_001166597.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034626.mm.1 |
NM_138306.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034626.mm.1 |
NM_138306.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034626.mm.1 |
TC0200004130.mm.1 |
MTA TC |