Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epsti1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029495.2 | NP_083771.1 | 9 - 10 | 1005 | 69 | |
GenBank mRNA | AK077223.1 | - | 9 - 10 | 811 | 69 | |
AK077376.1 | - | 9 - 10 | 840 | 69 | ||
AK135710.1 | - | 9 - 10 | 999 | 69 | ||
AK136055.1 | - | 9 - 10 | 1006 | 69 | ||
AY239224.1 | - | 9 - 10 | 911 | 69 | ||
BC020120.1 | - | 9 - 10 | 838 | 3988503 | 69 | |
BC021821.1 | - | 9 - 10 | 865 | 3581940 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |