Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pwp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029546.2 | NP_083822.1 | 12 - 13 | 1545 | 66 | |
GenBank mRNA | AB041855.1 | - | 12 - 13 | 1506 | 66 | |
AK088254.1 | - | 12 - 13 | 1546 | 66 | ||
AK088983.1 | - | 12 - 13 | 1546 | 66 | ||
BC112325.1 | - | 12 - 13 | 1511 | 5355810 | 66 | |
BC128480.1 | - | 12 - 13 | 1400 | 40097069 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |