Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sun5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK006225.1 | - | 7 - 8 | 524 | 128 | |
AY307077.1 | - | 7 - 8 | 543 | 128 | ||
FJ667498.1 | - | 8 - 9 | 532 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |