Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bag6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252468.1 | NP_001239397.1 | 20 - 21 | 3227 | 73 | |
NM_001252469.1 | NP_001239398.1 | 21 - 22 | 3138 | 73 | ||
NM_057171.2 | NP_476512.1 | 21 - 22 | 3281 | 73 | ||
GenBank mRNA | AK148796.1 | - | 20 - 21 | 3147 | 73 | |
AK164266.1 | - | 21 - 22 | 3111 | 73 | ||
AK170134.1 | - | 22 - 23 | 3206 | 73 | ||
AK182155.1 | - | 2 - 3 | 197 | 73 | ||
BC026647.1 | - | 13 - 14 | 2022 | 73 | ||
BC030296.1 | - | 2 - 3 | 150 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |