Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scaf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134123.3 | NP_598884.2 | 15 - 16 | 2206 | 71 | |
GenBank mRNA | AK032418.1 | - | 408 | 71 | ||
AK122451.1 | - | 15 - 16 | 2073 | 71 | ||
AK161113.1 | - | 1 - 2 | 118 | 71 | ||
BC030846.1 | - | 2 - 3 | 204 | 71 | ||
BC038363.1 | - | 15 - 16 | 1850 | 5371946 | 71 | |
BC075621.1 | - | 15 - 16 | 2199 | 6406441 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |