Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scaf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134123.3 | NP_598884.2 | 5 - 6 | 893 | 57 | |
GenBank mRNA | AK122451.1 | - | 5 - 6 | 760 | 57 | |
AK171078.1 | - | 6 - 7 | 700 | 57 | ||
AK200783.1 | - | 1 - 2 | 73 | 57 | ||
AK218201.1 | - | 1 - 2 | 73 | 57 | ||
BC038363.1 | - | 5 - 6 | 537 | 5371946 | 57 | |
BC075621.1 | - | 5 - 6 | 886 | 6406441 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |