Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iws1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173441.3 | NP_775617.1 | 2 - 3 | 417 | 69 | |
GenBank mRNA | AK028247.1 | - | 2 - 3 | 388 | 69 | |
AK033013.1 | - | 2 - 3 | 412 | 69 | ||
AK041691.1 | - | 2 - 3 | 411 | 69 | ||
AK163972.1 | - | 2 - 3 | 388 | 69 | ||
BC068184.1 | - | 2 - 3 | 357 | 30639737 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |