Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175022.2 | NP_778187.2 | 10 - 11 | 5622 | 118 | |
GenBank mRNA | AK032894.1 | - | 7 - 8 | 2110 | 118 | |
AK173130.1 | - | 7 - 8 | 5061 | 118 | ||
BC025846.1 | - | 2 - 3 | 783 | 118 | ||
BC058674.1 | - | 7 - 8 | 4261 | 118 | ||
BC075628.1 | - | 11 - 12 | 4532 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |