Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hapln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178255.3 | NP_839986.2 | 1 - 2 | 111 | 69 | |
GenBank mRNA | AK016123.1 | - | 1 - 2 | 112 | 69 | |
AK017865.1 | - | 1 - 2 | 90 | 69 | ||
AK136956.1 | - | 103 | 69 | |||
AY262757.1 | - | 1 - 2 | 111 | 69 | ||
AY269788.1 | - | 1 - 2 | 53 | 69 | ||
BC110474.2 | - | 1 - 2 | 83 | 40045754 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |