Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thnsl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033929.2 | NP_001029101.1 | 3 - 4 | 519 | 76 | |
NM_178413.5 | NP_848500.3 | 4 - 5 | 587 | 76 | ||
GenBank mRNA | AK165463.1 | - | 3 - 4 | 485 | 76 | |
BC025604.1 | - | 4 - 5 | 550 | 5250319 | 76 | |
BC051244.1 | - | 3 - 4 | 481 | 4975661 | 76 | |
CB600010.1 | - | 4 - 5 | 557 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |