Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199247.1 | NP_001186176.1 | 6 - 7 | 502 | 74 | |
NM_001199248.1 | NP_001186177.1 | 5 - 6 | 601 | 74 | ||
NM_001199249.1 | NP_001186178.1 | 5 - 6 | 601 | 74 | ||
NM_178637.2 | NP_848752.1 | 7 - 8 | 658 | 74 | ||
NR_037603.1 | 4 - 5 | 534 | 74 | |||
GenBank mRNA | AB055409.1 | - | 4 - 5 | 512 | 74 | |
AF528194.1 | - | 3 - 4 | 212 | 74 | ||
AF528195.1 | - | 6 - 7 | 593 | 74 | ||
AF528196.1 | - | 5 - 6 | 437 | 74 | ||
AK143674.1 | - | 5 - 6 | 553 | 74 | ||
AK146352.1 | - | 7 - 8 | 615 | 74 | ||
AK159568.1 | - | 5 - 6 | 534 | 74 | ||
AK160077.1 | - | 5 - 6 | 534 | 74 | ||
BC110675.1 | - | 7 - 8 | 599 | 4011689 | 74 | |
BC129967.1 | - | 5 - 6 | 436 | 74 | ||
BC129968.1 | - | 5 - 6 | 437 | 40106268 | 74 | |
DQ139980.1 | - | 6 - 7 | 593 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |