Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tiparp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178892.5 | NP_849223.2 | 2 - 3 | 1148 | 142 | |
GenBank mRNA | AK028529.1 | - | 2 - 3 | 1149 | 142 | |
AK032453.1 | - | 2 - 3 | 1132 | 142 | ||
AK150248.1 | - | 2 - 3 | 1130 | 142 | ||
AK162094.1 | - | 2 - 3 | 1154 | 142 | ||
AK163347.1 | - | 2 - 3 | 1188 | 142 | ||
BC003919.1 | - | 1 - 2 | 408 | 142 | ||
BC068173.1 | - | 2 - 3 | 1122 | 30616032 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |