Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181517.3 | NP_852658.2 | 12 - 13 | 1329 | 111 | |
GenBank mRNA | AK039534.1 | - | 12 - 13 | 1481 | 111 | |
AY316153.1 | - | 12 - 13 | 1329 | 111 | ||
BC053524.1 | - | 9 - 10 | 1012 | 111 | ||
BC064825.1 | - | 3 - 4 | 285 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |