Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016891.3 | NP_058587.1 | 3 - 4 | 327 | 104 | |
GenBank mRNA | AB021743.1 | - | 3 - 4 | 275 | 104 | |
AK054239.1 | - | 3 - 4 | 287 | 104 | ||
AK078135.1 | - | 3 - 4 | 306 | 104 | ||
AK088776.1 | - | 3 - 4 | 299 | 104 | ||
BC006606.1 | - | 3 - 4 | 327 | 3157293 | 104 | |
BC052678.1 | - | 3 - 4 | 323 | 30062580 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Mus musculus Mouse Wnt Panel 1700 Inventoried Probe spans exons |