Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vasp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282021.1 | NP_001268950.1 | 10 - 11 | 1275 | 66 | |
NM_001282022.1 | NP_001268951.1 | 10 - 11 | 1248 | 66 | ||
NM_009499.3 | NP_033525.2 | 10 - 11 | 1278 | 66 | ||
NR_104069.1 | 8 - 9 | 839 | 66 | |||
GenBank mRNA | AF084548.1 | - | 10 - 11 | 1245 | 66 | |
AK140329.1 | - | 10 - 11 | 1242 | 66 | ||
AK150103.1 | - | 10 - 11 | 1240 | 66 | ||
AK155480.1 | - | 10 - 11 | 1241 | 66 | ||
AK157021.1 | - | 10 - 11 | 1239 | 66 | ||
AK170780.1 | - | 10 - 11 | 1204 | 66 | ||
AK171302.1 | - | 10 - 11 | 1243 | 66 | ||
AK171715.1 | - | 10 - 11 | 1242 | 66 | ||
BC010223.1 | - | 10 - 11 | 980 | 3500587 | 66 | |
BC015289.1 | - | 10 - 11 | 1221 | 4240907 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |