Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301737.1 | NP_001288666.1 | 4 - 5 | 716 | 78 | |
NM_010065.3 | NP_034195.2 | 4 - 5 | 716 | 78 | ||
NR_125959.1 | 4 - 5 | 716 | 78 | |||
GenBank mRNA | AK011651.1 | - | 4 - 5 | 683 | 78 | |
AK144142.1 | - | 4 - 5 | 667 | 78 | ||
AK220483.1 | - | 4 - 5 | 658 | 78 | ||
BC034679.1 | - | 4 - 5 | 670 | 4506865 | 78 | |
BC058623.1 | - | 4 - 5 | 650 | 5706803 | 78 | |
CA328097.1 | - | 2 - 3 | 342 | 78 | ||
L31395.1 | - | 4 - 5 | 613 | 78 | ||
L31397.1 | - | 4 - 5 | 669 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |