Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136070.1 | NP_001129542.1 | 5 - 6 | 694 | 80 | |
NM_010709.3 | NP_034839.2 | 5 - 6 | 694 | 80 | ||
GenBank mRNA | AK030312.1 | - | 5 - 6 | 80 | ||
AK034115.1 | - | 5 - 6 | 666 | 80 | ||
AK036154.1 | - | 5 - 6 | 690 | 80 | ||
AK046539.1 | - | 5 - 6 | 706 | 80 | ||
AK047005.1 | - | 5 - 6 | 689 | 80 | ||
AK082748.1 | - | 5 - 6 | 709 | 80 | ||
AK083638.1 | - | 5 - 6 | 654 | 80 | ||
AK139052.1 | - | 5 - 6 | 694 | 80 | ||
AK161269.1 | - | 5 - 6 | 677 | 80 | ||
AK166085.1 | - | 5 - 6 | 706 | 80 | ||
AK167980.1 | - | 5 - 6 | 690 | 80 | ||
AK168120.1 | - | 5 - 6 | 660 | 80 | ||
AK170252.1 | - | 5 - 6 | 721 | 80 | ||
AK170448.1 | - | 5 - 6 | 706 | 80 | ||
AK171115.1 | - | 5 - 6 | 672 | 80 | ||
BC025036.1 | - | 5 - 6 | 650 | 4935223 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |