Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pstpip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011193.2 | NP_035323.2 | 2 - 3 | 569 | 77 | |
GenBank mRNA | AK037936.1 | - | 2 - 3 | 571 | 77 | |
AK088150.1 | - | 2 - 3 | 566 | 77 | ||
BC096761.1 | - | 2 - 3 | 349 | 30040955 | 77 | |
U87814.1 | - | 2 - 3 | 409 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |