Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rusc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083807.1 | NP_001077276.1 | 5 - 6 | 1872 | 61 | |
NM_001083808.1 | NP_001077277.1 | 4 - 5 | 412 | 61 | ||
NM_028188.2 | NP_082464.2 | 6 - 7 | 2283 | 61 | ||
GenBank mRNA | AK032334.1 | - | 4 - 5 | 902 | 61 | |
AK039664.1 | - | 6 - 7 | 2211 | 61 | ||
AK042689.1 | - | 5 - 6 | 1820 | 61 | ||
AK170012.1 | - | 4 - 5 | 426 | 61 | ||
AK170210.1 | - | 4 - 5 | 375 | 61 | ||
BC056360.1 | - | 5 - 6 | 1810 | 6816267 | 61 | |
BC057034.1 | - | 4 - 5 | 478 | 5687517 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |