Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081256.1 | NP_001074725.1 | 20 - 21 | 4874 | 98 | |
GenBank mRNA | AK033343.1 | - | 6 - 7 | 798 | 98 | |
AK159090.1 | - | 7 - 8 | 1208 | 98 | ||
BC031981.1 | - | 6 - 7 | 943 | 98 | ||
BC038376.1 | - | 10 - 11 | 1692 | 98 | ||
BC060727.1 | - | 13 - 14 | 2434 | 98 | ||
BC108415.1 | - | 13 - 14 | 2278 | 98 | ||
BC151084.1 | - | 20 - 21 | 4835 | 9087997 | 98 | |
BC172150.1 | - | 19 - 20 | 4195 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |