Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177320.2 | NP_796294.2 | 2 - 3 | 252 | 56 | |
GenBank mRNA | AK089348.1 | - | 2 - 3 | 253 | 56 | |
AK089483.1 | - | 2 - 3 | 257 | 56 | ||
AK149847.1 | - | 2 - 3 | 220 | 56 | ||
AK154643.1 | - | 2 - 3 | 229 | 56 | ||
AK171279.1 | - | 2 - 3 | 253 | 56 | ||
AY156924.1 | - | 1 - 2 | 101 | 56 | ||
BC128076.1 | - | 1 - 2 | 101 | 40060132 | 56 | |
BC128077.1 | - | 1 - 2 | 101 | 40060133 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |