Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rftn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181397.2 | NP_852062.1 | 8 - 9 | 1543 | 58 | |
GenBank mRNA | AK082517.1 | - | 1156 | 58 | ||
AK087803.1 | - | 1317 | 58 | |||
AK088466.1 | - | 6 - 7 | 1080 | 58 | ||
AK152130.1 | - | 8 - 9 | 1465 | 58 | ||
AK153288.1 | - | 8 - 9 | 1465 | 58 | ||
AK155870.1 | - | 8 - 9 | 1507 | 58 | ||
AK172884.1 | - | 8 - 9 | 1471 | 58 | ||
BC043324.1 | - | 4 - 5 | 600 | 58 | ||
BC052074.1 | - | 8 - 9 | 1543 | 6401192 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |