Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmiz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183208.3 | NP_899031.2 | 6 - 7 | 891 | 72 | |
GenBank mRNA | AK054366.1 | - | 6 - 7 | 785 | 72 | |
BC058646.1 | - | 6 - 7 | 809 | 6405777 | 72 | |
BC065120.1 | - | 6 - 7 | 784 | 6856060 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |