Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spink2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289764.1 | NP_001276693.1 | 2 - 3 | 125 | 83 | |
NM_001289767.1 | NP_001276696.1 | 2 - 3 | 221 | 83 | ||
NM_183284.3 | NP_899107.1 | 2 - 3 | 204 | 83 | ||
GenBank mRNA | AK005709.1 | - | 2 - 3 | 187 | 83 | |
BC087876.1 | - | 2 - 3 | 167 | 6773100 | 83 | |
DV068802.1 | - | 2 - 3 | 224 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |