Search Thermo Fisher Scientific
Gene Symbol: Rims4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183023.1 | NP_898844.1 | 3 - 4 | 352 | 81 | |
GenBank mRNA | AK134613.1 | - | 3 - 4 | 598 | 81 | |
AY326955.1 | - | 3 - 4 | 352 | 81 | ||
BC115475.1 | - | 3 - 4 | 351 | 81 | ||
BC115476.1 | - | 3 - 4 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |