Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnlip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026925.3 | NP_081201.2 | 5 - 6 | 487 | 77 | |
GenBank mRNA | AY387690.1 | - | 4 - 5 | 459 | 77 | |
BC061061.1 | - | 5 - 6 | 487 | 30316303 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |