Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900008712.mm.1 |
NM_001199247.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008712.mm.1 |
NM_001199247.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008712.mm.1 |
NM_001199248.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008712.mm.1 |
NM_001199248.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008712.mm.1 |
NM_001199249.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008712.mm.1 |
NM_001199249.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008712.mm.1 |
NM_178637.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008712.mm.1 |
NM_178637.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008712.mm.1 |
TC1900000975.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900008708.mm.1 |
NM_001199247.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008708.mm.1 |
NM_001199247.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008708.mm.1 |
NM_001199248.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008708.mm.1 |
NM_001199248.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008708.mm.1 |
NM_001199249.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008708.mm.1 |
NM_001199249.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008708.mm.1 |
NM_178637.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008708.mm.1 |
NM_178637.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008708.mm.1 |
TC1900000975.mm.1 |
MTA TC |