Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007713.4 | NP_031739.3 | 5 - 6 | 1316 | 63 | |
GenBank mRNA | AF033565.1 | - | 5 - 6 | 592 | 63 | |
AK009908.1 | - | 1 - 2 | 35 | 63 | ||
AK090215.1 | - | 5 - 6 | 557 | 63 | ||
AK133936.1 | - | 5 - 6 | 562 | 63 | ||
AK136481.1 | - | 3 - 4 | 2955 | 63 | ||
AK140015.1 | - | 4 - 5 | 2659 | 63 | ||
AK146942.1 | - | 5 - 6 | 1319 | 63 | ||
AK154003.1 | - | 5 - 6 | 565 | 63 | ||
AK167300.1 | - | 5 - 6 | 1318 | 63 | ||
BC085084.1 | - | 5 - 6 | 559 | 6309020 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |