Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mboat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153546.4 | NP_705774.1 | 1 - 2 | 276 | 118 | |
GenBank mRNA | AB297382.1 | - | 1 - 2 | 99 | 118 | |
AB305045.1 | - | 1 - 2 | 99 | 118 | ||
AK033388.1 | - | 1 - 2 | 218 | 118 | ||
AK137755.1 | - | 1 - 2 | 208 | 118 | ||
AK146984.1 | - | 1 - 2 | 183 | 118 | ||
BC023845.1 | - | 1 - 2 | 280 | 5346000 | 118 | |
BC024653.1 | - | 1 - 2 | 228 | 3660479 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |