Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109988.1 | NP_001103458.1 | 3 - 4 | 285 | 75 | |
NM_001109989.2 | NP_001103459.1 | 3 - 4 | 208 | 75 | ||
NM_001267717.1 | NP_001254646.1 | 3 - 4 | 197 | 75 | ||
NM_053078.4 | NP_444308.1 | 2 - 3 | 323 | 75 | ||
GenBank mRNA | AK020629.1 | - | 3 - 4 | 146 | 75 | |
AK048361.1 | - | 2 - 3 | 123 | 75 | ||
AK081502.1 | - | 2 - 3 | 124 | 75 | ||
AK133769.1 | - | 1 - 2 | 1418 | 75 | ||
BC054762.1 | - | 2 - 3 | 105 | 6414312 | 75 | |
BQ715224.1 | - | 2 - 3 | 110 | 75 | ||
BU057200.1 | - | 2 - 3 | 113 | 75 | ||
CJ148521.1 | - | 2 - 3 | 428 | 75 | ||
CJ165393.1 | - | 3 - 4 | 288 | 75 | ||
CX768079.1 | - | 3 - 4 | 206 | 75 | ||
EU447302.1 | - | 1 - 2 | 51 | 75 | ||
X70398.1 | - | 2 - 3 | 119 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |