Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phtf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172992.3 | NP_766580.3 | 7 - 8 | 867 | 81 | |
GenBank mRNA | AK035536.1 | - | 7 - 8 | 821 | 81 | |
AK037021.1 | - | 7 - 8 | 754 | 81 | ||
AK042382.1 | - | 7 - 8 | 846 | 81 | ||
AK163376.1 | - | 7 - 8 | 793 | 81 | ||
BC052841.1 | - | 7 - 8 | 780 | 30050876 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |